Skip to Content
Merck
All Photos(1)

Key Documents

860467P

Avanti

N-C16-desoxymethylsphingosine

Avanti Research - A Croda Brand 860467P, powder

Synonym(s):

N-palmitoyl-1-desoxymethylsphingosine (m17:1/16:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H65NO2
CAS Number:
Molecular Weight:
507.87
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860467P-1mg)
pkg of 1 × 5 mg (860467P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860467P

lipid type

sphingolipids
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)CNC(CCCCCCCCCCCCCCC)=O

InChI key

RMAWKILAWRQSBQ-XBSMHFKOSA-N

General description

Desoxymethylsphingosine is an atypical deoxy-sphingoid base.

Application

N-C16-desoxymethylsphingosine or N-palmitoyl-1-desoxymethylsphingosine (m17:1/16:0) has been used as a standard in the quantitation of  atypical sphingoid bases in biological samples by reverse-phase liquid chromatography coupled to electrospray ionization tandem mass spectrometry (LC-MS/MS).

Packaging

5 mL Amber Glass Screw Cap Vial (860467P-1mg)
5 mL Amber Glass Screw Cap Vial (860467P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacques Bodennec et al.
Journal of lipid research, 44(7), 1413-1419 (2003-05-06)
We describe a new method that permits quantification in the pmol to nmol range of three lyso-neutral glycosphingolipids (lyso-n-GSLs), glucosylsphingosine (GlcSph), galactosylsphingosine (GalSph), and lactosylsphingosine, in the same sample as neutral glycosphingolipids (n-GSLs). Lyso-n-GSLs and n-GSLs are initially obtained from
Junliang Wan et al.
Journal of agricultural and food chemistry, 67(46), 12953-12961 (2019-10-23)
Most common sphingolipids are comprised of "typical" sphingoid bases (sphinganine, sphingosine, and structurally related compounds) and are produced via the condensation of l-serine with a fatty acyl-CoA by serine palmitoyltransferase. Some organisms, including mammals, also produce "atypical" sphingoid bases that
Shu-Chih Chen-Quay et al.
Journal of pharmaceutical sciences, 98(2), 606-619 (2008-06-20)
Tight junctions (TJs) play an important role in regulating paracellular drug transport. The aim of this study was to identify lipids that rapidly and reversibly alter transepithelial electrical resistance (TER) and/or TJ permeability in epithelial tissue. In this study, we
Terina N Martinez et al.
Molecular neurodegeneration, 7, 45-45 (2012-09-15)
Dopaminergic (DA) neurons in the ventral midbrain selectively degenerate in Parkinson's disease (PD) in part because their oxidative environment in the substantia nigra (SN) may render them vulnerable to neuroinflammatory stimuli. Chronic inhibition of soluble Tumor Necrosis Factor (TNF) with
Sarah T Pruett et al.
Journal of lipid research, 49(8), 1621-1639 (2008-05-24)
"Sphingosin" was first described by J. L. W. Thudichum in 1884 and structurally characterized as 2S,3R,4E-2-aminooctadec-4-ene-1,3-diol in 1947 by Herb Carter, who also proposed the designation of "lipides derived from sphingosine as sphingolipides." This category of amino alcohols is now

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service