Skip to Content
Merck
All Photos(2)

Key Documents

850524P

Avanti

CHEMS

Avanti Research - A Croda Brand

Synonym(s):

3β-Hydroxy-5-cholestene 3-hemisuccinate; 5-Cholesten-3β-ol 3-hemisuccinate; CHS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H50O4
CAS Number:
Molecular Weight:
486.73
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25

description

cholesteryl hemisuccinate

Assay

>99% (TLC)

form

powder

mol wt

486.73 g/mol

packaging

pkg of 1 × 1 g (850524P-1g)
pkg of 1 × 5 g (850524P-5g)

manufacturer/tradename

Avanti Research - A Croda Brand

technique(s)

protein quantification: suitable

application(s)

cell analysis

shipped in

dry ice

storage temp.

−20°C

SMILES string

O([C@H]1CC[C@@]2([C@@H]3[C@H]([C@H]4[C@@]([C@H](CC4)[C@@H](CCCC(C)C)C)(CC3)C)CC=C2C1)C)C(=O)CCC(=O)O

InChI

1S/C31H50O4/c1-20(2)7-6-8-21(3)25-11-12-26-24-10-9-22-19-23(35-29(34)14-13-28(32)33)15-17-30(22,4)27(24)16-18-31(25,26)5/h9,20-21,23-27H,6-8,10-19H2,1-5H3,(H,32,33)/t21-,23+,24+,25-,26+,27+,30+,31-/m1/s1

InChI key

WLNARFZDISHUGS-MIXBDBMTSA-N

General description

Cholesteryl hemisuccinate (CHEMS) is an acidic cholesterol ester.

Application

CHEMS is suitable:
  • to prepare multifunctional envelope-type nano device (MEND) by a lipid film hydration method based on electrostatic interactions
  • to as a component in bolavesicle
  • to prepare liposome (LS)
  • for the preparation of anionic liposomes by the lipid thin film hydration and freeze/thaw technique and to encapsulate ovalbumin (OVA) and listeriolysin O (LLO)
  • to prepare liposomal membranes to study its effect on the physical–chemical properties of 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine (POPC) liposomes

Biochem/physiol Actions

CHEMS can help to maintain the stability of dioleoylphosphatidylethanolamine (DOPE). This amphipathic lipid is used in mixtures with dioleoylphosphatidylethanolamine (DOPE) to form ′pH sensitive′ fusogenic vesicles. It can control cell growth.

Packaging

20 mL Clear Glass Screw Cap Vial (850524P-1g)
30 mL Amber Wide Mouth Glass Bottle with Screw Cap (850524P-5g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipogels for Encapsulation of Hydrophilic Proteins and Hydrophobic Small Molecules
Homyak CC, et al.
Biomacromolecules, 19(1), 132-140 (2017)
Effect of cholesteryl hemisuccinate on the interfacial properties of phosphatidylcholine bilayers
Massey JB
Biochimica et Biophysica Acta - Biomembranes, 1415(1), 193-204 (1998)
Yongrong Yao et al.
Biomaterials science, 8(1), 256-265 (2019-11-07)
Chemotherapy is a dominant treatment modality for different types and stages of cancer. However, hypoxia is one of the undesirable limitations of chemotherapy, which reduces the therapeutic efficiency in cancer treatment, ultimately leading to failure of the treatment. Herein, an
Enhanced in vivo gene expression mediated by listeriolysin O incorporated anionic LPDII: Its utility in cytotoxic T lymphocyte-inducing DNA vaccine
Sun X, et al.
J. Controlled Release, 148(2), 219-225 (2010)
Preparation and evaluation of multifunctional envelope-type nano device loading siRNA
Yin H, et al.
Artificial Cells, Nanomedicine, and Biotechnology (Print), 44(1), 20-26 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service