Skip to Content
Merck
All Photos(2)

Key Documents

850456P

Avanti

16:0-18:0 PC

Avanti Research - A Croda Brand

Synonym(s):

1-hexadecanoyl-2-octadecanoyl-sn-glycero-3-phosphocholine; PSPC; PC(16:0/18:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H84NO8P
CAS Number:
Molecular Weight:
762.09
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (850456P-1g)
pkg of 1 × 25 mg (850456P-25mg)
pkg of 1 × 500 mg (850456P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

InChI

1S/C42H84NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h40H,6-39H2,1-5H3/t40-/m1/s1

InChI key

PZNPLUBHRSSFHT-RRHRGVEJSA-N

General description

Phosphatidylcholines (PCs) are phospholipids, that are abundantly present in the tissues of the animal. 16:0-18:0 PC (PSPC/1-palmitoyl-2-stearoyl-sn-glycero-3-phosphocholine) is a saturated lipid , which consists of two additional methylene units in the sn-2 acyl chain.

Application

16:0-18:0 PC (1-palmitoyl-2-stearoyl-sn-glycero-3-phosphocholine) has been used in liposome preparation. It may be used as a phospholipid standard.

Biochem/physiol Actions

Phosphatidylcholines (PCs) play a functional role in reducing the level of blood lipid and cholesterol.

Packaging

20 mL Clear Glass Screw Cap Vial (850456P-1g)
20 mL Clear Glass Screw Cap Vial (850456P-500mg)
5 mL Amber Glass Screw Cap Vial (850456P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipids II: Phospholipids, Glycosphingolipids, and Cholesterol
Essentials of Medical Biochemistry, 299-320 (2015)
German Stepanov et al.
FEBS letters, 583(1), 97-100 (2008-12-09)
We attempted to evaluate the affinity of the anionic phospholipids to cytochrome c by means of surface plasmon resonance (SPR) technique and to correlate it with the cytochrome c active site alterations and peroxidase activity. Our experiments showed a strong
Joseph A Hankin et al.
Journal of the American Society for Mass Spectrometry, 18(9), 1646-1652 (2007-07-31)
Common organic matrix-assisted laser desorption/ionization (MALDI) matrices, 2,5-dihydroxybenzoic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, and alpha-cyano-4-hydroxycinnamic acid, were found to undergo sublimation without decomposition under conditions of reduced pressure and elevated temperature. This solid to vapor-phase transition was exploited to apply MALDI matrix
Miao-Miao Zhou et al.
Lipids in health and disease, 15(1), 135-135 (2016-08-26)
Phosphatidylcholine (PC), the major source of dietary choline, has been demonstrated to improve the capability of learning and memory in rodent and the amelioration of long-chain n-3 polyunsaturated fatty acids (PUFA) on anti-aging and anti-oxidation is widely known as well.
Simon Becher et al.
Analytical chemistry, 90(19), 11486-11494 (2018-09-11)
Phosphatidylcholines are the major phospholipid component of most eukaryotic cell membranes. Phosphatidylcholines have been shown to actively participate in regulatory and metabolic processes. Dysfunctional metabolic processes have been linked to human disease and can result in altered phosphatidylcholine structural features

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service