Skip to Content
Merck
All Photos(2)

Key Documents

850315P

Avanti

08:0 PC

1,2-dioctanoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

PC(8:0/8:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H48NO8P
CAS Number:
Molecular Weight:
509.61
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 200 mg (850315P-200mg)
pkg of 1 × 25 mg (850315P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850315P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC)=O)COC(CCCCCCC)=O)=O

General description

Phosphatidylcholine (PC) is one of the most important constituent of the mucosal layer of the colon. 08:0 PC (1,2-dioctanoyl-sn-glycero-3-phosphocholine/C8PC) is a water soluble short alkyl chain lipid.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

08:0 PC (1,2-dioctanoyl-sn-glycero-3-phosphocholine/C8PC) has been used as a substrate to study the analytical capabilities of several methods to determine water soluble phospholipids containing choline (wCh-PL) and also to study the behavior of the its analytical signal in a synthetic serum matrix (data-Trol). It may also be used as a bicellar model to study their phase behavior and to form a lipid film to study its effect on P-glycoprotein′s (P-gp′s) activity/to compare its effect with TWEEN®80 and Cremophor®EL.

Biochem/physiol Actions

Phosphatidylcholine (PC) can form a hydrophobic surface in the mucus by acting as a surfactant to inhibit the penetrance of bacteria.

Packaging

20 mL Clear Glass Screw Cap Vial (850315P-200mg)
5 mL Clear Glass Sealed Ampule (850315P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Cremophor is a registered trademark of BASF SE
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phase behavior of a binary lipid system containing long- and short-chain phosphatidylcholines
Sun HY, et al.
Royal Society of Chemistry Advances, 7(10), 5715-5724 null
Manuel Weinheimer et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 108, 13-22 (2016-09-04)
The efflux transporter P-glycoprotein (P-gp) significantly modulates drug transport across the intestinal mucosa, strongly reducing the systemic absorption of various active pharmaceutical ingredients. P-gp inhibitors could serve as helpful tools to enhance the oral bioavailability of those substances. As a
Isabel Sanz-Vicente et al.
Analytical biochemistry, 519, 30-37 (2016-12-14)
In this paper we present methods to determine water soluble phospholipids containing choline (wCh-PL). The analytes were hydrolyzed by the enzyme phospholipase D and the choline formed was oxidized by the enzyme Choline Oxidase (ChOx); the fluorescence changes of the
Inflammatory Bowel Disease
Integrative Medicine, 501-516 null
Teruhiko Baba et al.
Chemistry and physics of lipids, 227, 104870-104870 (2020-01-10)
Interfacial properties and membrane protein solubilization activity of a series of partially fluorinated octyl-phosphocholine (PC) surfactants were investigated from the viewpoint of the fluorination degree of the hydrophobic chain. The critical micelle concentration (CMC), surface tension lowering activity, molecular occupied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service