Skip to Content
Merck
All Photos(2)

Key Documents

850312P

Avanti

DOCPe

Avanti Research - A Croda Brand 850312P, powder

Synonym(s):

2-((2,3-bis(oleoyloxy)propyl)dimethylammonio)ethyl ethyl phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H86NO8P
CAS Number:
Molecular Weight:
800.14
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 200 mg (850312P-200mg)
pkg of 1 × 25 mg (850312P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850312P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(C[N+](C)(CCOP([O-])(OCC)=O)C)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C45H86NO8P/c1-6-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-44(47)51-42-43(41-46(4,5)39-40-53-55(49,50)52-8-3)54-45(48)38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-7-2/h21-24,43H,6-20,25-42H2,1-5H3/b23-21-,24-22-

InChI key

WDTDXEZJIUAULM-SXAUZNKPSA-N

General description

2-((2,3-bis(oleoyloxy)propyl)dimethylammonio)ethyl ethylphosphate (DOCPe) is a neutrally charged lipid.
Inverse-phosphocholine lipids, a remix of a common phospholipid, contain headgroups with an inverted charge orientation relative to phosphocholine (PC) lipids. The iPC lipid headgroup has a quaternary amine adjacent to the bilayer interface and a phosphate that extends into the aqueous phase. Therefore iPC lipids afford a unique opportunity to investigate the biophysical and bioactivity-related ramifications of a charge inversion at the bilayer surface.

Application

DOCPe is suitable for liposomes preparation.

Packaging

20 mL Clear Glass Screw Cap Vial (850312P-200mg)
5 mL Clear Glass Sealed Ampule (850312P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charge and Coordination Directed Liposome Fusion onto SiO2 and TiO2 Nanoparticles
Wang X, et al.
Langmuir, 35(5), 1672-1681 (2018)
Emily K Perttu et al.
Journal of the American Chemical Society, 134(10), 4485-4488 (2012-03-01)
Zwitterionic inverse-phosphocholine (iPC) lipids contain headgroups with an inverted charge orientation relative to phosphocholine (PC) lipids. The iPC lipid headgroup has a quaternary amine adjacent to the bilayer interface and a phosphate that extends into the aqueous phase. Neutral iPC
Feng Wang et al.
Journal of the American Chemical Society, 137(36), 11736-11742 (2015-08-25)
Zwitterionic phosphocholine (PC) lipids are highly biocompatible, representing a major component of the cell membrane. A simple mixing of PC liposomes and silica (SiO2) surface results in liposome fusion with the surface and formation of supported lipid bilayers. However, the
Yibo Liu et al.
Langmuir : the ACS journal of surfaces and colloids, 34(32), 9337-9348 (2018-03-13)
Phospholipids are a major component of the cell membrane. In most natural phospholipids, the phosphate acts as a bridge, connecting the other portion of the polar headgroup with the hydrophobic tails. Such bridging phosphate is chemically quite inert. Synthetic lipids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service