Skip to Content
Merck
All Photos(2)

Key Documents

850176P

Avanti

06:0 PI(3,4,5)P3

1,2-dihexanoyl-sn-glycero-3-phospho-(1′-myo-inositol-3′,4′,5′-trisphosphate) (ammonium salt), powder

Synonym(s):

PIP3[3′,4′,5′](6:0/6:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H54N4O22P4
CAS Number:
Molecular Weight:
838.56
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (850176P-1MG)
pkg of 1 × 100 μg (with stopper and crimp cap (850176P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850176P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 850176P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP(O[C@H]1[C@H](O)[C@@H](OP([O-])(O)=O)[C@H](OP(O)([O-])=O)[C@@H](OP(O)([O-])=O)[C@H]1O)([O-])=O)(OC(CCCCC)=O)COC(CCCCC)=O.[NH4+].[NH4+].[NH4+].[NH4+]

InChI key

YTZPLJSHLDNARZ-KPKFNREXSA-N

General description

Inositol phospholipid species are membrane-bound signaling molecules that have been implicated in almost all aspects of cellular physiology, including cellular growth, metabolism, proliferation, and survival.

Biochem/physiol Actions

Phosphatidylinositol 3,4,5-trisphosphate (PI(3,4,5)P3) acts as a lipid second messenger. It is capable of triggering protein kinase B (PKB) phosphorylation. PI(3,4,5) may play a role in the modulation of synaptic activity.

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850176P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850176P-500ug)
5 mL Amber Glass Screw Cap Vial (850176P-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rays H Y Jiang et al.
Cell host & microbe, 11(2), 99-100 (2012-02-22)
Survival of blood stage malaria parasites requires extensive host cell remodeling, which is facilitated by secretion of parasite proteins via a dedicated protein export pathway. In a recent Cell paper, Bhattacharjee et al., (2012) describe PI(3)P binding as one of the
Sulochanadevi Baskaran et al.
Molecular cell, 47(3), 339-348 (2012-06-19)
Macroautophagy is essential to cell survival during starvation and proceeds by the growth of a double-membraned phagophore, which engulfs cytosol and other substrates. The synthesis and recognition of the lipid phosphatidylinositol 3-phosphate, PI(3)P, is essential for autophagy. The key autophagic
Kewei Ma et al.
Cellular signalling, 20(4), 684-694 (2008-02-06)
The PI3K-PKB pathway is an important and widely studied pathway in cell signaling. The enzyme activity of PI3K produces D-3 phosphoinositides, including the lipid second messengers PI(3,4,5)P3 and PI(3,4)P2. PI(3,4,5)P3 has been deemed to be the most important second messenger
Thang Manh Khuong et al.
Neuron, 77(6), 1097-1108 (2013-03-26)
PI(3,4,5)P3 is a low-abundance lipid thought to play a role in the regulation of synaptic activity; however, the mechanism remains obscure. We have constructed novel split Venus-based probes and used superresolution imaging to localize PI(3,4,5)P3 at Drosophila larval neuromuscular synapses. We find
Gerald R V Hammond et al.
Science (New York, N.Y.), 337(6095), 727-730 (2012-06-23)
The quantitatively minor phospholipid phosphatidylinositol (4,5)-bisphosphate [PI(4,5)P(2)] fulfills many cellular functions in the plasma membrane (PM), whereas its synthetic precursor, phosphatidylinositol 4-phosphate (PI4P), has no assigned PM roles apart from PI(4,5)P(2) synthesis. We used a combination of pharmacological and chemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service