Skip to Content
Merck
All Photos(2)

Key Documents

850115P

Avanti

IP3(1,4,5)

Avanti Research - A Croda Brand

Synonym(s):

D-myo-inositol-1,4,5-triphosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H24N3O15P3
CAS Number:
Molecular Weight:
471.19
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (850115P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850115P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H]([C@H](O)[C@H](O)[C@@H](OP(O)([O-])=O)[C@@H]1OP([O-])(O)=O)OP(O)([O-])=O.[NH4+].[NH4+].[NH4+]

Application

IP3(1,4,5) has been used as a supplement to treat purified inositol 1,4,5-trisphosphate 3-kinase A (IP3K-A) recombinant protein in kinase-Glo luminescent kinase assay.

Biochem/physiol Actions

Inositol 1,4,5-triphosphate (IP3) acts as a second messenger. IP3 is capable of liberating calcium from the endoplasmic reticulum. IP3 helps to modulate the interaction between IP3R-binding protein released with inositol 1,4,5-trisphosphate (IRBIT) and inositol 1,4,5-trisphosphate receptor (IP3R).

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850115P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850115P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hideaki Ando et al.
The Journal of biological chemistry, 278(12), 10602-10612 (2003-01-15)
The inositol 1,4,5-trisphosphate (IP(3)) receptors (IP(3)Rs) are IP(3)-gated Ca(2+) channels on intracellular Ca(2+) stores. Herein, we report a novel protein, termed IRBIT (IP(3)R binding protein released with inositol 1,4,5-trisphosphate), which interacts with type 1 IP(3)R (IP(3)R1) and was released upon
Spinocerebellar ataxia type 14
Handbook of Clinical Neurophysiology, 103, 555-559 null
Jack D Thatcher
Science signaling, 3(119), tr3-tr3 (2010-04-29)
This resource provides and describes two animated lessons that illustrate inositol 1,4,5-trisphosphate signal transduction pathways. They are designed for any collegiate-level course that presents the basic steps of the prototypical pathway. Courses that might employ these animations include Introductory Biology
Dongmin Lee et al.
Protein expression and purification, 97, 72-80 (2014-03-01)
Inositol 1,4,5-trisphosphate 3-kinase A (IP3K-A, alternative name: ITPKA) is a neuron-specific enzyme that converts 1,4,5-trisphosphate (IP3) into inositol 1,3,4,5-tetrakisphosphate (IP4) through its kinase domain. In addition, transient overexpression of IP3K-A induces morphological changes in dendritic spines of excitatory synapses in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service