Skip to Content
Merck
All Photos(1)

Key Documents

700184M

Avanti

desmosterol (D6) ester

26,26,26,27,27,27-hexadeuteriocholesta-5,24-dien-3β-ol (9Z-octadecenoate), methylene chloride solution

Synonym(s):

111014

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H70D6O2
CAS Number:
Molecular Weight:
655.12
UNSPSC Code:
12352211
NACRES:
NA.12

Assay

>99% (TLC)

form

methylene chloride solution

packaging

pkg of 1 × 1 mL (700184M-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700184M

concentration

1 mg/mL (700184M-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

Desmosterol is a precursor of cholesterol synthesis and the enzyme 24-dehydrocholesterol reductase catalyzes this step. It lacks a double bond between C24 and C25, which is present in cholesterol. Desmosterol is the major brain sterol and is localized in spermatozoa membrane and astrocytes. Desmosterol (D6) ester is a deuterated desmosterol with octadecanoic acid.

Biochem/physiol Actions

Desmosterol is a liver X receptor (LXR) agonist and it regulates inflammatory signaling in macrophages. Desmosterol accumulates during impairment of 24-dehydrocholesterol reductase enzyme thereby leading to desmosterolosis. It is a potential therapeutic target for treating the neurodegenerative disorder.

Packaging

5 mL Clear Glass Sealed Ampule (700184M-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saara Vainio et al.
The Journal of biological chemistry, 281(1), 348-355 (2005-10-27)
Desmosterol is an immediate precursor of cholesterol in the Bloch pathway of sterol synthesis and an abundant membrane lipid in specific cell types. The significance of the difference between the two sterols, an additional double bond at position C24 in
Maurice Jansen et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(3), 865-870 (2012-12-12)
The brain is the most cholesterol-enriched tissue in the body. During brain development, desmosterol, an immediate precursor of cholesterol, transiently accumulates up to 30% of total brain sterols. This massive desmosterol deposition appears to be present in all mammalian species
Phillip A Wages et al.
Journal of lipid research, 59(10), 1916-1926 (2018-08-09)
Regulating blood cholesterol (Chol) levels by pharmacotherapy has successfully improved cardiovascular health. There is growing interest in the role of Chol precursors in the treatment of diseases. One sterol precursor, desmosterol (Des), is a potential pharmacological target for inflammatory and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service