700146P
Avanti
alkyne-cholesterol
27-norcholest-5-en-25-yn-3β-ol, powder
Synonym(s):
alkynyl cholesterol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C26H40O
CAS Number:
Molecular Weight:
368.60
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Recommended Products
Assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 100 μg (700146P-100ug)
pkg of 1 × 250 μg (700146P-250ug)
manufacturer/tradename
Avanti Research™ - A Croda Brand 700146P
lipid type
click reagents
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
Alkyne-cholesterol is a clickable cholesterol probe proprietary for Avanti Polar Lipids. They differ from cholesterol by having an alkyne group.
This reagent is a modified lipid containing an omega-terminal alkyne. The terminal alkyne group can be used in a highly specific linking reaction with azide-containing reagents, known as ‘click chemistry′, in the presence of a copper (Cu)-containing catalyst.
Biochem/physiol Actions
Alkyne-cholesterol is not applicable for use in living cells due to copper ion (Cu (I) and Cu (II)) requirements. It aids in easy and sensitive detection of cholesterol using various biophysical methods. Alkyne-cholesterol can be conjugated to boron-dipyrromethene (BODIPY) fluorescent dye. It is useful in cholesterol trafficking studies in various organelles including mitochondria, endoplasmic reticulum and Golgi.
Packaging
5 mL Amber Glass Screw Cap Vial (700146P-100ug)
5 mL Amber Glass Screw Cap Vial (700146P-250ug)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Cholesterol: Revisiting its fluorescent journey on 200th anniversary of Chevruel?s ?cholesterine?
Chaudhuri A and Anand D
Biomedical Spectroscopy and Imaging , 6(1-2), 1/24/2020-1/24/2020 (2017)
J Králová et al.
Folia biologica, 65(1), 1-10 (2019-06-07)
Cellular cholesterol plays fundamental and diverse roles in many biological processes and affects the pathology of various diseases. Comprehensive and detailed understanding of the cellular functions and characteristics of cholesterol requires visualization of its subcellular distribution, which can be achieved
Katarzyna A Solanko et al.
Lipid insights, 8(Suppl 1), 95-114 (2015-01-01)
Cholesterol transport between cellular organelles comprised vesicular trafficking and nonvesicular exchange; these processes are often studied by quantitative fluorescence microscopy. A major challenge for using this approach is producing analogs of cholesterol with suitable brightness and structural and chemical properties
Katherine Windsor et al.
Journal of lipid research, 54(10), 2842-2850 (2013-07-06)
Lipid modifications aid in regulating (and misregulating) protein function and localization. However, efficient methods to screen for a lipid's ability to modify proteins are not readily available. We present a strategy to identify protein-reactive lipids and apply it to a
Kristina Hofmann et al.
Journal of lipid research, 55(3), 583-591 (2013-12-18)
Cholesterol is an important lipid of mammalian cells and plays a fundamental role in many biological processes. Its concentration in the various cellular membranes differs and is tightly regulated. Here, we present a novel alkyne cholesterol analog suitable for tracing
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service