Skip to Content
Merck
All Photos(2)

Key Documents

700016P

Avanti

cholesterol sulfate

Avanti Research - A Croda Brand 700016P

Synonym(s):

cholesterol sulfonate; cholesteryl sulfate; sodium cholesteryl sulfate; cholesteryl sulphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H45NaO4S
CAS Number:
Molecular Weight:
488.70
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

description

cholesterol 3-sulfate (sodium salt)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (700016P-1g)
pkg of 1 × 25 mg (700016P-25mg)
pkg of 1 × 500 mg (700016P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700016P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[S](=O)(=O)([O-])O[C@H]1CC[C@@]2([C@@H]3[C@H]([C@H]4[C@@]([C@H](CC4)[C@@H](CCCC(C)C)C)(CC3)C)CC=C2C1)C

InChI

1S/C27H46O4S.Na/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(31-32(28,29)30)13-15-26(20,4)25(22)14-16-27(23,24)5;/h9,18-19,21-25H,6-8,10-17H2,1-5H3,(H,28,29,30);/q;+1/p-1/t19-,21+,22+,23-,24+,25+,26+,27-;/m1./s1

InChI key

LMPVQXVJTZWENW-KPNWGBFJSA-M

General description

Cholesterol sulfate is synthesized by the sulfoconjugation of cholesterol in the presence of enzyme cholesterol sulfotransferase. It is present in brain, lungs, hair and skin tissues as well as cells including erythrocytes, platelets and spermatozoa.

Application

Cholesterol sulfate has been used in micelle preparation and in surface plasmon resonance (SPR) studies.

Biochem/physiol Actions

Cholesterol sulfate is associated with atherosclerotic lesions and serves as a substrate for platelet adhesion. It stabilizes the spermatozoa membranes and erythrocytes. Cholesterol sulfate is involved in the keratinocyte differentiation and its levels are elevated in recessive X-linked ichthyosis. It serves as a ligand for retinoic acid-related orphan receptor α (RORα). Cholesterol sulfate regulates glucose metabolism and is a potential target for metabolic disorders. It is also a potential marker for human uterine cervical carcinoma tissue and prostate carcinoma.

Packaging

20 mL Clear Glass Screw Cap Vial (700016P-1g)
20 mL Clear Glass Screw Cap Vial (700016P-500mg)
5 mL Amber Glass Screw Cap Vial (700016P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles A Strott et al.
Journal of lipid research, 44(7), 1268-1278 (2003-05-06)
Cholesterol sulfate is quantitatively the most important known sterol sulfate in human plasma, where it is present in a concentration that overlaps that of the other abundant circulating steroid sulfate, dehydroepiandrosterone (DHEA) sulfate. Although these sulfolipids have similar production and
M Merten et al.
Circulation, 103(16), 2032-2034 (2001-04-25)
Cholesterol 3-sulfate is present on a variety of cells and in human LDL, and it has been found in atherosclerotic lesions of human aorta. Its precise biological role has not yet been described. In this study, we investigated the interaction
Patrizia Lopalco et al.
Frontiers in physiology, 10, 1344-1344 (2019-11-19)
The reduction of sperm motility and count, or oligoasthenozoospermia, is one of the major causes of reduced fertility or infertility in men. Lipid composition of spermatozoa is important in determining their functional characteristics, in particular on motility, acrosomal exocytosis or
Xiongjie Shi et al.
Molecular and cellular biology, 34(3), 485-497 (2013-11-28)
Sulfotransferase (SULT)-mediated sulfation represents a critical mechanism in regulating the chemical and functional homeostasis of endogenous and exogenous molecules. The cholesterol sulfotransferase SULT2B1b catalyzes the sulfoconjugation of cholesterol to synthesize cholesterol sulfate (CS). In this study, we showed that the
Alexey V Kostarnoy et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(13), E2758-E2765 (2017-03-16)
Sterile (noninfected) inflammation underlies the pathogenesis of many widespread diseases, such as allergies and autoimmune diseases. The evolutionarily conserved innate immune system is considered to play a key role in tissue injury recognition and the subsequent development of sterile inflammation;

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service