Skip to Content
Merck
All Photos(1)

Key Documents

131101P

Avanti

Brain Extract Total

Avanti Research - A Croda Brand

Synonym(s):

Brain Total Lipid Extract (Porcine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 1 g (131101P-1g)
pkg of 1 × 100 mg (131101P-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

lipid extracts

shipped in

dry ice

storage temp.

−20°C

General description

Total Lipid Extract is a chloroform:methanol extract of the respective tissue (Brain, Soybean, Heart, etc.). This extract is partitioned against deionized water, and the chloroform phase is concentrated.

Application

Brain extract total has been used for the preparation of liposomes.[1] It has also been used as a component of lipid cocktail for lipid cosedimentation assay.[2]

Packaging

5 mL Clear Glass Sealed Ampule (131101P-100mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (131101P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and characterization of Rvs162/Rvs167-3, a novel N-BAR heterodimer in the human fungal pathogen Candida albicans
Gkourtsa A, et al.
Eukaryotic Cell, 14(2), 182-193 (2015)
Single particle fluorescence burst analysis of epsin induced membrane fission
Brooks A, et al.
Testing, 10(3), e0119563-e0119563 (2015)
Ziyi Yang et al.
Bioorganic & medicinal chemistry, 28(7), 115374-115374 (2020-02-25)
A series of novel flurbiprofen-clioquinol hybrids were designed and synthesized as multifunctional agents for Alzheimer's disease therapy, and their potential was evaluated through various biological experiments. In vitro studies showed that most target compounds exhibited significant ability to inhibit self-
Wei Li et al.
Bioorganic chemistry, 97, 103707-103707 (2020-03-09)
A series of pyridoxine-resveratrol hybrids were designed and synthesized as monoamine oxidase B inhibitors for the treatment of Parkinson's disease. Most of them exhibited potent inhibitory activities on MAO-B with high selectivity. Specifically, compounds 12a, 12g and 12l showed the
B Sandrino et al.
Chemistry and physics of lipids, 230, 104930-104930 (2020-05-30)
Understanding protein aggregation is essential to unveil molecular mechanisms associated with neurodegenerative diseases such as Alzheimer's, Huntington's and spongiform encephalopathy, particularly to determine the role of interaction with cell membranes. In this study, we employ Langmuir monolayers as cell membrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service