Z685356
Platinum dish, with reinforced rim
capacity 100 mL
Synonym(s):
Dish for High-Temperature Use, Platinum Lab Dish, Reinforced Platinum Dish
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41122200
NACRES:
NB.43
Recommended Products
material
platinum
packaging
pack of 1 ea
manufacturer/tradename
Birmingham metals BD9
H × diam.
29 mm × 75.0 mm
capacity
100 mL
dish diam.
75 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Platinum dish, with reinforced rim is made of precious metals that play an important role in the chemical analysis of materials used for high temperature applications.
Platinum labware has excellent resistance to acids such as acetic, hydrochloric, hydrofluoric, hydroiodic, nitric, selenic, and sulfuric, and to oxidation at high temperatures. Dishes of platinum can be cleaned by boiling in a suitable solvent. For the removal of substances which have alloyed with the surface, scour with alumina powder.
Features and Benefits
- Primarily made from platinum.
- Can also be fabricated using 10% Rh-Pt., 5% Au-Pt., pure Pt., Ag., and Au.
- Supplied without a pouring lip unless otherwise specified.
- Manufactured to a tolerance of +/-5% of the weight.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service