material
polypropylene
feature
Holds 12 x 0.5 mL tubes
Holds 12 x 8 mm tubes
Holds 32 x 0.2 mL tubes
Holds 32 x 6.5 mm tubes
Holds 6 x 15 mL tubes
Holds 6 x 16.4 mm tubes
Holds 9 x 1.5-2 mL tubes
Holds 9 x 11.2 mm tubes
Microtubes
manufacturer/tradename
Heathrow Scientific HS29040B
size
210x100x100 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service