Z675474
Sterile economy sample spoon
8 mL
Synonym(s):
dispensing tool
All Photos(1)
About This Item
UNSPSC Code:
41122403
NACRES:
NB.35
Recommended Products
description
PS
manufacturer/tradename
Bel-Art F36740-00004
Related Categories
General description
Moulded of lightweight and rigid, white high impact polystyrene. Individually packaged sterile spoons for one time use packed 100 per box.
- Use to collect specific volumes of dry or liquid materials
- Remains upright when filled or empty for use as a weighing boat
Choose from one of the most recent versions:
Certificates of Analysis (COA)