Key Documents
Z563099
J-KEM® thermocouple probe
Single-element, thermocouple type, T, diam. × L 1/8 in. × 6 in.
About This Item
UNSPSC Code:
41112303
NACRES:
NB.76
Pricing and availability is not currently available.
description
PTFE-coated
feature
thermocouple type T
manufacturer/tradename
J-Kem
diam. × L
1/8 in. × 6 in.
Legal Information
J-KEM is a registered trademark of J-KEM Electronics, Inc.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. what is the connector like? flat or round prong?
    1 answer
    1. The connector has round pins, Type OST.
      Helpful?