Z400432
Ace pressure tubes with plunger valves and thermowells
capacity ~100 mL
About This Item
Recommended Products
material
PTFE cap
borosilicate glass tube
cap (Front-Seal Plunger Valve)
clear tube
glass
round bottom
screw top cap (plugseal)
white cap
sterility
non-sterile
feature
graduations: no
internal thread
packaging
pack of 1 ea
manufacturer/tradename
Ace Glass 8648-170
L × O.D.
17.8 cm × 38.1 mm
capacity
~100 mL
diam.
38.1 mm
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Complete item consists of tube with #15 Ace-Thred, PTFE bushing with FETFE® O-ring, and plunger valve with thermowell.
Replacement FETFE O-rings:
Ace-Thred #Front SealBack Seal
7 Z504661 Z504742
15 Z504858 Z505021
25 Z505056 Z505110
CAUTION: This glassware is not guaranteed against breakage caused by pressure or vacuum. These tubes should never be used if scratched or otherwise damaged.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service