Key Documents
Z275344
Sigma-Aldrich
Cannon-Fenske viscometer tube
size 300
About This Item
UNSPSC Code:
41111700
NACRES:
PB.36
size
300
description
constant ~ 0.25 cSt/s
manufacturer/tradename
Ace Glass 798845
viscosity measuring range
50-250 cSt
Related Categories
General description
Obtain kinematic viscosities using tubes designed for ASTM Method D445 and D2515. Viscosities can be measured with a precision of ±0.2% quickly and easily.
Requires approximately 7 mL of sample. Certificate of Calibration supplied.
Choose from one of the most recent versions:
Certificates of Analysis (COA)