size
2
packaging
pkg of 12 ea
H × thickness
21 mm × 2.5 mm
bottom O.D. × I.D.
16 mm × 11 mm
top O.D. × I.D.
27 mm × 22 mm
General description
For Buchner, conical, fritted-glass, and porcelain filter funnels. Gray neoprene rubber adapters fit singly or can be nested around funnel stem to produce a good fit. Withstands autoclaving.
Choose from one of the most recent versions:
Certificates of Analysis (COA)