Key Documents
Z225096
Sigma-Aldrich
Scienceware® mini-vacuum desiccator
pkg of 1 ea
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
Pricing and availability is not currently available.
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F41099-0000
I.D. × H
4 in. × 2 7/16 in.
General description
The Scienceware mini-vacuum desiccator has a clear molded polycarbonate top which snap locks to the bottom polypropylene plate against a Buna-N O-ring. A polypropylene stopcock with 2 mm bore PTFE plug permits slow release of vacuum. Vacuum rating is 740 mm Hg.
Not compatible with organic solvents and vapors.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Srishti Dar et al.
Nature protocols, 12(2), 390-400 (2017-01-27)
The process of membrane fission is fundamental to diverse cellular processes such as nutrient uptake, synaptic transmission and organelle biogenesis, and it involves the localized application of curvature stress to a tubular membrane intermediate, forcing it to undergo scission. Alternative
Questions
1–5 of 5 Questions  
  1. What's the internal diameter of the dessiccator?
    1 answer
    1. As listed on the Product Detail Page in the PROPERTIES section, the product dimensions are 4 in. × 2 7/16 in. ( I.D. × H.). The weight of the desiccator is 19oz.
      Helpful?
  2. Can the product Z225096 be subjected to a temperature of 140°C for a duration of 2 hours?
    1 answer
    1. The Scienceware mini-vacuum desiccator Z225096 cannot be heated as it would melt, according to the supplier.
      Helpful?
  3. Do the Scienceware® brand polypropylene products contain any leachable material?
    1 answer
    1. Although Bel-Art does not specifically test their Scienceware® brand polypropylene products, they have no evidence of any problems with leachable material in the polypropylene.
      Helpful?
  4. What is the Department of Transportation shipping information for this product?
    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
      Helpful?
  5. Potete fornirmi dimensioni e peso del mini essiccatore a vuoto?
    0 answers