Skip to Content
Merck
All Photos(1)

Key Documents

Z177369

Scienceware® micro funnel

top I.D. 35 mm, stem diam. 8 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121809
NACRES:
NB.43

packaging

pack of 12 ea

manufacturer/tradename

Bel-Art F14685-0035

stem diam.

8 mm

top I.D.

35 mm

Looking for similar products? Visit Product Comparison Guide

General description

PP. Useful for micro work, filling burettes, manometers, etc.

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service