Key Documents
Z154253
Sigma-Aldrich
Aldrich® double-chamber vacuum trap
W × H 97 mm × 250 mm
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
W × H
97 mm × 250 mm
General description
Inlet and outlet have serrated fittings, 14 to 16 mm. Can be used in a 2 L Dewar flask.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC