Skip to Content
Merck
All Photos(1)

Documents

W515302

Sigma-Aldrich

2-Ethylhexanoic acid

≥99%

Synonym(s):

2-Ethylcaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(C2H5)CO2H
CAS Number:
Molecular Weight:
144.21
Beilstein:
1750468
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.078
NACRES:
NA.21

biological source

synthetic

vapor density

4.98 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)
10 mmHg ( 115 °C)

Assay

≥99%

autoignition temp.

699 °F

expl. lim.

1.04 %, 135 °F
8.64 %, 188 °F

refractive index

n20/D 1.425 (lit.)

bp

228 °C (lit.)

density

0.903 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cucumber

SMILES string

CCCCC(CC)C(O)=O

InChI

1S/C8H16O2/c1-3-5-6-7(4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)

InChI key

OBETXYAYXDNJHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

237.2 °F - closed cup

Flash Point(C)

114 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shashank Mishra et al.
Chemical Society reviews, 36(11), 1770-1787 (2008-01-25)
This critical review deals with the chemistry and applications of metal alkanoates with medium size (C5 to C12) carbon chain length. A particular emphasis is given to metal 2-ethylhexanoates, which find wide applications as metal-organic precursors in materials science, as
R Gaudin et al.
International archives of occupational and environmental health, 84(5), 523-531 (2010-08-31)
The aim of this study was to assess, by biological monitoring, workers' exposure to di(2-ethylhexyl) phthalate (DEHP) in the flexible-PVC industry in France to provide additional occupational exposure data, which are particularly scarce. Over 5 days of pre-and post-shift sampling
Yan Xu et al.
Archives of toxicology, 80(5), 293-298 (2005-12-06)
The highly directional maternal-to-fetal transfer of essential fatty acids (EFAs) across the placenta plays a critical role in guiding proper fetal development. Exposure to xenobiotics that may alter the fetal supply of EFAs/lipids could lead to fetal toxicity. Since the
S Elss et al.
Food additives and contaminants, 21(8), 811-814 (2004-09-17)
After simultaneous distillation-extraction (SDE) of commercial baby foods (n = 20) and fruit juices (n = 15) (among them 15 and eight products labelled 'organic', respectively) from 11 different suppliers, analyses performed by coupled capillary gas chromatography-mass spectrometry (GC-MS) revealed
R Gaudin et al.
International archives of occupational and environmental health, 81(8), 959-966 (2007-12-22)
The aim of this study was to perform a biological monitoring survey of workers exposed to di(2-ethylhexyl)phthalate (DEHP) in a factory using polyvinyl chloride plastisols and to contribute additional occupational data of exposure particularly sparse in the industrial sectors where

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service