Skip to Content
Merck
All Photos(1)

Key Documents

W366706

Sigma-Aldrich

Diphenyl ether

≥99%, FG

Synonym(s):

Diphenyl oxide, Phenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2O
CAS Number:
Molecular Weight:
170.21
FEMA Number:
3667
Beilstein:
1364620
EC Number:
Council of Europe no.:
2201
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.035
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

vapor density

>5.86 (25 °C, vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥99%

form

solid or liquid

autoignition temp.

1144 °F

expl. lim.

1.5 %

refractive index

n20/D 1.579 (lit.)

bp

259 °C (lit.)

mp

25-27 °C (lit.)

density

1.073 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

geranium; green

SMILES string

O(c1ccccc1)c2ccccc2

InChI

1S/C12H10O/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10H

InChI key

USIUVYZYUHIAEV-UHFFFAOYSA-N

Gene Information

human ... TTR(7276)

Looking for similar products? Visit Product Comparison Guide

General description

Diphenyl ether is used as a flavor additive in tobacco products such as bidi cigarettes.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of flavor-related compounds in the unburned contents of bidi and clove cigarettes.
Stanfill SB, et al.
Journal of Agricultural and Food Chemistry, 54(22), 8580-8588 (2006)
Quantitation of ten flavor compounds in unburned tobacco products.
Lisko JG, et al.
Analytical Methods : Advancing Methods and Applications, 6(13), 4698-4704 (2014)
Ariana Beste et al.
The Journal of organic chemistry, 74(7), 2837-2841 (2009-03-06)
Lignin is an abundant natural resource that is a potential source of valuable chemicals. Improved understanding of the pyrolysis of lignin occurs through the study of model compounds for which phenethyl phenyl ether (PhCH(2)CH(2)OPh, PPE) is the simplest example representing
Ge-Fei Hao et al.
Journal of computer-aided molecular design, 25(3), 213-222 (2011-01-25)
Protoporphyrinogen oxidase (PPO, EC 1.3.3.4), which has been identified as a significant target for a great family of herbicides with diverse chemical structures, is the last common enzyme responsible for the seventh step in the biosynthetic pathway to heme and
Takayuki Yonezawa et al.
Bioorganic & medicinal chemistry letters, 21(11), 3248-3251 (2011-05-10)
Osteogenic activity of six diarylheptanoids, acerogenin A (1), (R)-acerogenin B (2), aceroside I (3), aceroside B(1) (4), aceroside III (5) and (-)-centrolobol (6) and two phenolic compounds; (+)-rhododendrol (7) and (+)-cathechin (8), isolated from the stem bark of Acer nikoense

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service