Skip to Content
Merck
All Photos(1)

Key Documents

W350206

Sigma-Aldrich

2-Mercapto-3-butanol

mixture of isomers, ≥97%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH(SH)CH3
CAS Number:
Molecular Weight:
106.19
FEMA Number:
3502
EC Number:
Council of Europe no.:
760
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.024
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥97%

refractive index

n20/D 1.48 (lit.)

bp

53 °C/10 mmHg (lit.)

density

1.013 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty; alliaceous; sulfurous

SMILES string

CC(O)C(C)S

InChI

1S/C4H10OS/c1-3(5)4(2)6/h3-6H,1-2H3

InChI key

MJQWABQELVFQJL-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

Taste at 4 ppm

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 21, 52-52 (1996)
Zemfira S Kinzyabaeva et al.
Ultrasonics sonochemistry, 67, 105169-105169 (2020-05-18)
We have discovered a selective and efficient method for the synthesis of previously unknown 1,9-(1',4'-oxathiano)-1,9-dihydro-(C60-Ih)[5,6]fullerene, a compound with the direct attachment of the sulfur atom to the fullerene core. The method is based on the reaction of C60 with 1,2-hydroxythiols

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service