Skip to Content
Merck
All Photos(1)

Key Documents

W338818

Sigma-Aldrich

2-Tridecanone

≥96%, FG

Synonym(s):

Methyl undecyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10COCH3
CAS Number:
Molecular Weight:
198.34
FEMA Number:
3388
Beilstein:
1757402
EC Number:
Council of Europe no.:
11194
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.103
NACRES:
NA.21
Organoleptic:
coconut; dairy; fatty; waxy; earthy
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥96%

refractive index

n20/D 1.435 (lit.)

bp

134 °C/10 mmHg (lit.)
263 °C (lit.)

mp

24-27 °C (lit.)

density

0.822 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

coconut; dairy; fatty; waxy; earthy

SMILES string

CCCCCCCCCCCC(C)=O

InChI

1S/C13H26O/c1-3-4-5-6-7-8-9-10-11-12-13(2)14/h3-12H2,1-2H3

InChI key

CYIFVRUOHKNECG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Tridecanone occurs naturally in the glandular trichomes of Lycopersicon hirsutum f. glabratum, a wild tomato species. This allelochemical is responsible for its insecticidal resistance against Lepidopterous larvae and Heliothis zea.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Toxicity studies of analogs of 2-tridecanone, a naturally occurring toxicant from a wild tomato.
Dimock MB, et al.
Journal of Chemical Ecology, 8(5), 837-842 (1982)
2-Tridecanone: a naturally occurring insecticide from the wild tomato Lycopersicon hirsutum f. glabratum
Williams WG, et al.
Science, 207(4433), 888-889 (1980)
2?tridecanone, tomatoes and Heliothis zea: potential incompatibility of plant antibiosis with insecticidal control.
Kennedy GG, et al.
Entomologia Experimentalis et Applicata, 35(3), 305-311 (1984)
Persistence of 2-tridecanone on the leaves of seven vegetables.
G F Antonious
Bulletin of environmental contamination and toxicology, 73(6), 1086-1093 (2005-01-28)
P J Skiba et al.
Insect biochemistry and molecular biology, 24(8), 847-853 (1994-09-01)
A fatty acid chain elongation process is involved in incorporation of saturated and unsaturated fatty acyl-CoA esters into 2-tridecanone and (Z)-10-heptadecen-2-one by Drosophila buzzatii. The microsomal fraction from mature male ejaculatory bulbs is chain-length specific and requires malonyl-CoA (or acetyl-CoA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service