Skip to Content
Merck
All Photos(1)

Key Documents

W335809

Sigma-Aldrich

2-Methoxy-3(5 or 6)-isopropylpyrazine

≥98%, FG

Synonym(s):

3,5 Or 6-Isopropyl-2-methoxypyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2O
CAS Number:
Molecular Weight:
152.19
FEMA Number:
3358
EC Number:
Council of Europe no.:
11344
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.121
NACRES:
NA.21
Organoleptic:
green; earthy; pea; pepper
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

refractive index

n20/D 1.495 (lit.)

density

0.996 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; earthy; pea; pepper

SMILES string

COc1cnc(cn1)C(C)C.COc2cncc(n2)C(C)C.COc3nccnc3C(C)C

InChI

1S/3C8H12N2O/c1-6(2)7-4-10-8(11-3)5-9-7;1-6(2)7-4-9-5-8(10-7)11-3;1-6(2)7-8(11-3)10-5-4-9-7/h3*4-6H,1-3H3

InChI key

MQHZLANELJKBBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methoxy-3(5or 6)-isopropylpyrazine is a mixture of positional isomers commonly used as a flavoring agent in the food industry. Pyrazine derivatives exhibit a wide variety of aromas in food. For instance, 2-methoxy-3-isopropyl pyrazine produces a green pea odor and earthy and beany flavors . In addition, they are naturally found in coffee beans, cocoa beans, nuts, and vegetables.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

152.6 °F

Flash Point(C)

67 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HS-SPME-GC-MS/olfactometry combined with chemometrics to assess the impact of germination on flavor attributes of chickpea, lentil, and yellow pea flours
Xu M, et al.
Food Chemistry, 280, 83-95 (2019)
The FEMA GRAS assessment of pyrazine derivatives used as flavor ingredients
Adams TB, et al.
Food And Chemical Toxicology, 40(4), 429-451 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service