W327700
Disodium succinate
≥98%
Synonym(s):
Succinic acid disodium salt, Sodium succinate dibasic
About This Item
Kosher
Recommended Products
biological source
synthetic
grade
Halal
Kosher
reg. compliance
FDA 21 CFR 117
Assay
≥98%
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
odorless
SMILES string
[Na+].[Na+].[O-]C(=O)CCC([O-])=O
InChI
1S/C4H6O4.2Na/c5-3(6)1-2-4(7)8;;/h1-2H2,(H,5,6)(H,7,8);;/q;2*+1/p-2
InChI key
ZDQYSKICYIVCPN-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Application of Electronic Tongue in Umami Detection and Soy Sauce Refining Process.: Utilizing disodium succinate, this paper discusses the application of electronic tongue technology in detecting umami flavors and refining soy sauce, highlighting innovations in food chemistry and sensory evaluation methods (Zhang et al., 2023).
- Fabrication of Succinate-Alginate Xerogel Films for In Vitro Coupling of Osteogenesis and Neovascularization.: This research explores the creation of succinate-alginate xerogel films aimed at enhancing osteogenesis and neovascularization in vitro, pointing to significant developments in biomaterials for tissue engineering (Deering et al., 2022).
Disclaimer
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service