Skip to Content
Merck
All Photos(1)

Key Documents

W284009

Sigma-Aldrich

ω-Pentadecalactone

≥98%, FG

Synonym(s):

Pentadecanolide, 1-Oxa-2-cyclohexadecanone, 15-Hydroxypentadecanoic acid lactone, 15-Pentadecanolide, Pentalide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H28O2
CAS Number:
Molecular Weight:
240.38
FEMA Number:
2840
Beilstein:
143710
EC Number:
Council of Europe no.:
181
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.004
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥98%

bp

137 °C/2 mmHg (lit.)

mp

34-38 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

animal; sweet

SMILES string

O=C1CCCCCCCCCCCCCCO1

InChI

1S/C15H28O2/c16-15-13-11-9-7-5-3-1-2-4-6-8-10-12-14-17-15/h1-14H2

InChI key

FKUPPRZPSYCDRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

ω-Pentadecalactone is a macrocyclic lactone that can be used as a flavoring agent and fragrance ingredient due to its musk flavor/odor. It occurs naturally in angelica root oil.

Application


  • Simple magnesium alkoxides: synthesis, molecular structure, and catalytic behaviour in the ring-opening polymerization of lactide and macrolactones and in the copolymerization of maleic anhydride and propylene oxide.: Examines the catalytic uses of ψ-pentadecalactone in polymer synthesis, highlighting its effectiveness in enhancing polymer properties and industrial applications (Wannipurage et al., 2023).

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyesters from Macrolactones Using Commercial Lipase NS 88011 and Novozym 435 as Biocatalysts.
Polloni AE, et al.
Applied Biochemistry and Biotechnology, 1-14 (2017)
Fragrance material review on ?-pentadecalactone.
McGinty D, et al.
Food And Chemical Toxicology, 49, S193-S201 (2011)
GRAS flavor chemicals-detection thresholds.
Leffingwell JC & Leffingwell, D.
Perfumer & Flavorist, 16(1), 1-19 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service