Skip to Content
Merck
All Photos(1)

Key Documents

W257508

Sigma-Aldrich

Hexyl octanoate

≥97%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CO2(CH2)5CH3
CAS Number:
Molecular Weight:
228.37
FEMA Number:
2575
EC Number:
Council of Europe no.:
394
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.113
NACRES:
NA.21
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.431 (lit.)

bp

307 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; fruity

SMILES string

CCCCCCCC(=O)OCCCCCC

InChI

1S/C14H28O2/c1-3-5-7-9-10-12-14(15)16-13-11-8-6-4-2/h3-13H2,1-2H3

InChI key

PBGWNXWNCSSXCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Najat H O Bahroun et al.
Analytica chimica acta, 1028, 121-130 (2018-06-10)
Rapid, sensitive, and selective detection and identification of pathogenic bacteria is required in terms of food security. In this study, exogenous VOCs liberated by Salmonella strains have been identified and quantified via head space-solid phase microextraction gas chromatography mass spectrometry
Elinor M Lichtenberg et al.
Behavioral ecology and sociobiology, 65(4), 763-774 (2011-04-09)
Foragers can improve search efficiency, and ultimately fitness, by using social information: cues and signals produced by other animals that indicate food location or quality. Social information use has been well studied in predator-prey systems, but its functioning within a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service