Skip to Content
Merck
All Photos(1)

Documents

W220108

Sigma-Aldrich

Butyl hexanoate

≥98%, FG

Synonym(s):

Butyl caproate, Hexanoic acid butyl ester, NSC 4022

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CO2(CH2)3CH3
CAS Number:
Molecular Weight:
172.26
FEMA Number:
2201
EC Number:
Council of Europe no.:
313
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.063
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.416 (lit.)

bp

61-62 °C/3 mmHg (lit.)

density

0.866 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

berry; fruity

SMILES string

CCCCCC(=O)OCCCC

InChI

1S/C10H20O2/c1-3-5-7-8-10(11)12-9-6-4-2/h3-9H2,1-2H3

InChI key

RPRPDTXKGSIXMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butyl hexanoate is an aliphatic ester that can be used as a flavoring agent. It is one of the main volatile compounds found in apples. Along with red sticky sphere traps, butyl hexanoate can also act as an odor lure for apple maggot flies.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fruit odor discrimination and sympatric host race formation in Rhagoletis
Linn C, et al
Proceedings of the National Academy of Sciences of the USA, 100(20), 11490-11493 (2003)
Evaluation of odor lures for use with red sticky spheres to trap apple maggot (Diptera: Tephritidae).
Reynolds AH & Prokopy RJ.
Journal of Economic Entomology, 90(6), 1655-1660 (1997)
M Guadalupe Sánchez-Otero et al.
Environmental technology, 31(10), 1101-1106 (2010-08-20)
The recombinant lipase LipMatCCR11 from the thermophilic strain Geobacillus thermoleovorans CCR11 was applied in the synthesis of n-butyl caproate via transesterification in hexane and xylene. The short chain flavour ester was obtained by alcoholysis from ethyl caproate and n-butyl alcohol
K Takahashi et al.
Biochemical and biophysical research communications, 131(2), 532-536 (1985-09-16)
Lipoprotein lipase was modified with 2,4-bis(O-methoxypolyethylene glycol)-6-chloro-s-triazine; forty-six percent out of seven amino groups in the molecule were substituted. The modified lipase catalyzed ester-exchange reactions between an ester and an alcohol, between an ester and an acid, and between two
Burdock GA
Encyclopedia of Food and Color Additives, 1, 341-342 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service