Skip to Content
Merck
All Photos(1)

Key Documents

T88595

Sigma-Aldrich

Trithiocyanuric acid

95%

Synonym(s):

1,3,5-Triazine-2,4,6-trithiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H3N3S3
CAS Number:
Molecular Weight:
177.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

>300 °C (lit.)

SMILES string

Sc1nc(S)nc(S)n1

InChI

1S/C3H3N3S3/c7-1-4-2(8)6-3(9)5-1/h(H3,4,5,6,7,8,9)

InChI key

WZRRRFSJFQTGGB-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeongmin Hong et al.
Journal of hazardous materials, 399, 123060-123060 (2020-09-18)
Heavy metal pollution caused by stormwater runoff has triggered a demand for effective heavy metal sorbents. Effective heavy metal removal using conventional stormwater runoff treatment processes that employ filtration mechanisms as primary removal mechanisms is difficult. Therefore, we attempt to
Xin Huang et al.
Chemosphere, 206, 513-521 (2018-05-20)
In this study, a novel magnetic nanoparticles (MNP) modified by an organodisulfide polymer (PTMT) was designed for adsorption of heavy metals (Hg(II), Pb(II) and Cd(II)) from simulated coal chemical high salinity wastewater. The MNP-PTMT nano-composite was synthesize and characterized by
Qi Zuo et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 34(12), 1405-1410 (2018-09-11)
A surface-enhanced Raman spectroscopy (SERS) based method was developed for the quantification of Cd2+ in rice. Gold nano-particles (AuNPs) modified with trimercaptotriazine served as a ratiometric SERS probe for the detection of Cd2+. A conical holed substrate was used to
Biing-Chiau Tzeng et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(71), 18033-18040 (2017-10-17)
Versatile building block [{Re(CO)
Duong Dinh Tuan et al.
Chemosphere, 261, 127552-127552 (2020-07-31)
As cobalt (Co) represents an effective transition metal for activating Oxone to degrade contaminants, tricobalt tetraoxide (Co3O4) is extensively employed as a heterogeneous phase of Co for Oxone activation. Since Co3O4 can be manipulated to exhibit various shapes, 2-dimensional plate-like

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service