Skip to Content
Merck
All Photos(1)

Documents

T28355

Sigma-Aldrich

N,N′-Diphenylthiourea

98%

Synonym(s):

1,3-Diphenyl-2-thiourea, Diphenylsulfourea, Thiocarbanilide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5NH)2CS
CAS Number:
Molecular Weight:
228.31
Beilstein:
644277
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

152-155 °C (lit.)

SMILES string

S=C(Nc1ccccc1)Nc2ccccc2

InChI

1S/C13H12N2S/c16-13(14-11-7-3-1-4-8-11)15-12-9-5-2-6-10-12/h1-10H,(H2,14,15,16)

InChI key

FCSHMCFRCYZTRQ-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2 - Skin Sens. 1 - STOT RE 2

Target Organs

Thyroid

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Contact allergy to IPPD and diphenylthiourea in an orthopaedic brace.
C G Aplin et al.
Contact dermatitis, 45(5), 301-302 (2001-11-28)
S Fregert et al.
Contact dermatitis, 8(1), 38-42 (1982-01-01)
28 patients reacting to a PVC adhesive tape used in routine patch testing gave positive reactions to diphenylthiourea (DPTU) and to phenylisothiocyanate (PITC) in low concentrations. DPTU is a heat stabilizer in the PVC and is partly decomposed to PITC.
Allergic contact dermatitis from diphenylthiourea in a wet suit.
W H Boehncke et al.
Contact dermatitis, 36(5), 271-271 (1997-05-01)
M Chen et al.
Electrophoresis, 15(10), 1290-1294 (1994-10-01)
Micellar capillary electrophoresis is optimized for separation of phenylthiohydantoin (PTH) amino acids produced in manual Edman degradation reaction for protein sequencing. There are also two major side-products produced by the Edman degradation reaction: diphenylthiourea and dimethylphenylthiourea. We report the complete
Graham A Bowmaker et al.
Dalton transactions (Cambridge, England : 2003), (39)(39), 8308-8316 (2009-10-01)
Crystallization of the copper(i) halides (CuX; X = Cl, Br, I) with N,N'-diphenylthiourea ('dptu' = (PhNH)(2)CS) in 1 : 2 ratio from acetonitrile solution in ambient conditions yields a mononuclear isomorphous series of complexes of the form [XCu(dptu)(2)].H(2)O, an unusually

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service