Key Documents
SYNV150500
Synthware photochemical reaction vessel with internal thread
500 mL
Synonym(s):
Synthware Double-Walled Quartz Photochemical Immersion Well,, Synthware V150500
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware V150500
capacity
500 mL
General description
Reaction Vessel constructed of borosilicate glass to accommodate immersion well. With #50 internal thread at top which accepts a RC50 bushing and Viton® O-ring to form a compression seal with the immersion well, one 14/20 joint for sparger tube, one 24/40 vertical joint for condenser and one #7 internal thread side arm for thermometer. Complete with quartz immersion well, caps and O-rings.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1