SYNS191530
Synthware™ metering valve stopcock plug
bore size 4 mm, Plug Size: 15.2/30
Synonym(s):
Synthware S191530
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S191530
bore size
4 mm
Related Categories
General description
With a threaded PTFE needle valve for the fine control of flow through the bore of the stopcock plug. This replacement kit is complete with all the components.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)