Documents
SYNF984500
Synthware Kuderna-Danish flask
capacity 500 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 19/22
Synonym(s):
Synthware F984500
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
pear-shaped
single-neck
Joint
bottom joint: ST/NS19/22
joint: ST/NS top24/40
Agency
suitable for EPA 1613
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F984500
capacity
500 mL
General description
Top joint is a 24/40 outer while the lower joint is a 19/22 inner.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Visit the Document Library
Related Content
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.