Key Documents
SYNF672460
Synthware pressure equalizing addition funnel with PTFE stopcock in equalizing line
60 mL, joint: ST/NS 24/40, stopcock: PTFE
Synonym(s):
Synthware F672460, addition funnel
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F672460
capacity
60 mL
60 mL
Related Categories
General description
With straight bore PTFE stopcock in pressure equalizing line, graduated.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)