Skip to Content
Merck
All Photos(1)

Key Documents

SYNF238530C

Synthware chromatography column with vacuum connection and fritted disk

joint: ST/NS 24/40, O.D. × L 90 mm × 300 mm, Disc porosity: coarse

Synonym(s):

Synthware F238530C, glass chromatography column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NB.51

Joint

joint: ST/NS24/40

grade

for chromatography

packaging

pkg of 1 ea

manufacturer/tradename

Synthware F238530C

O.D. × L

90 mm × 300 mm

Looking for similar products? Visit Product Comparison Guide

General description

Short column with lower 24/40 joint and hose connection for vacuum. All Synthware columns are heavy wall designed to increase safety.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service