Merck
SYNC264175
Synthware coil-style reflux condenser
joint: ST/NS 24/40, hose size 10 mm
Synonym(s):
Synthware C264175, coil-type condenser
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.51
UNSPSC Code:
41121800
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C264175
hose size
10 mm
General description
Coil style reflux condenser with tightly wrapped coil providing maximum cooling. Excellent for use with volatile solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint. Hose connections have an O.D. of 8mm or 10mm at the middle serration.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Global Trade Item Number
SKUGTIN
SYNC264175-1EA04061832186092