Merck
SYNC101075
Synthware Vigreux distillation column
joint: ST/NS 14/20, L 75 mm
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.51
UNSPSC Code:
41121800
Manufacturer/tradename:
Synthware C101075
Packaging:
pkg of 1 ea
Joint:
joint: ST/NS14/20
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C101075
L
75 mm
General description
Column length is measured from the highest to lowest indent in the column.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Global Trade Item Number
SKUGTIN
SYNC101075-1EA04061832185729