SLWSST3025
Pyrex® Stopcocks, plain arms
T-Key bore, PTFE key, bore 2.5 mm , key
Synonym(s):
Pyrex, SciLabware SST3025
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
material
PTFE key
manufacturer/tradename
SST3025
SciLabware
bore
2.5 mm , key
side arm O.D. × thickness
9 mm × 1.5 mm
stopcock diam.
14.5 mm
top diam.
14 mm
Looking for similar products? Visit Product Comparison Guide
General description
Borosilicate glass and PTFE liquid pathway
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service