Skip to Content
Merck
All Photos(3)

Key Documents

S7602

Sigma-Aldrich

Syringaldehyde

98%

Synonym(s):

3,5-Dimethoxy-4-hydroxybenzaldehyde, 4-Hydroxy-3,5-dimethoxybenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H2(OCH3)2CHO
CAS Number:
Molecular Weight:
182.17
Beilstein:
784514
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

bp

192-193 °C/14 mmHg (lit.)

mp

110-113 °C (lit.)

SMILES string

COc1cc(C=O)cc(OC)c1O

InChI

1S/C9H10O4/c1-12-7-3-6(5-10)4-8(13-2)9(7)11/h3-5,11H,1-2H3

InChI key

KCDXJAYRVLXPFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela Vieira Cortez et al.
Bioresource technology, 101(6), 1858-1865 (2009-10-14)
The effect of lignin degradation products liberated during chemical hydrolysis of lignocellulosic materials on xylose-to-xylitol bioconversion by Candida guilliermondii FTI 20037 was studied. Two aromatic aldehydes (vanillin and syringaldehyde) were selected as model compounds. A two-level factorial design was employed
Esteban D Babot et al.
Bioresource technology, 102(12), 6717-6722 (2011-04-23)
The ability of two natural phenols to act as mediators of the recombinant Myceliophthora thermophila laccase (MtL) in eucalypt-pulp delignification was investigated. After alkaline peroxide extraction, the properties of the enzymatically-treated pulps improved with respect to the control. The pulp
Rogério S Pereira et al.
Journal of industrial microbiology & biotechnology, 38(1), 71-78 (2010-09-08)
The inhibitory action of acetic acid, ferulic acid, and syringaldehyde on metabolism of Candida guilliermondii yeast during xylose to xylitol bioconversion was evaluated. Assays were performed in buffered and nonbuffered semidefined medium containing xylose as main sugar (80.0 g/l), supplemented
Chia-Hsin Huang et al.
Journal of natural products, 75(8), 1465-1468 (2012-08-14)
The antihyperglycemic effect of syringaldehyde (1), purified from the stems of Hibiscus taiwanensis, was investigated in streptozotocin-induced diabetic rats (STZ-diabetic rats) showing type-1 like diabetes mellitus. Bolus intravenous injection of 1 showed antihyperglycemic activity in a dose-dependent manner in STZ-diabetic
Palwinder Singh et al.
Journal of medicinal chemistry, 55(14), 6381-6390 (2012-06-28)
On the basis of structural analysis of dihydrofolate reductase (DHFR) (cocrystallized separately with NADPH, dihydrofolate and NADPH, trimethoprim), compounds 2 and 3 were optimized for inhibition of DHFR. Appreciable tumor growth inhibitory activities of compounds 2 and 3 over 60

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service