Skip to Content
Merck
All Photos(1)

Key Documents

P50919

Sigma-Aldrich

Propargylamine hydrochloride

95%

Synonym(s):

2-Propynylamine hydrochloride, 3-Amino-1-propyne hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CCH2NH2 · HCl
CAS Number:
Molecular Weight:
91.54
Beilstein:
3669858
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

powder

reaction suitability

reaction type: click chemistry

mp

179-182 °C (lit.)

storage temp.

2-8°C

SMILES string

Cl.NCC#C

InChI

1S/C3H5N.ClH/c1-2-3-4;/h1H,3-4H2;1H

InChI key

IKXNIQJDNKPPCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Hermanns et al.
Life science alliance, 3(9) (2020-07-29)
Deubiquitinating enzymes (DUBs) are important regulators of the posttranslational protein ubiquitination system. Mammalian genomes encode about 100 different DUBs, which can be grouped into seven different classes. Members of other DUB classes are found in pathogenic bacteria, which use them
Anna Fodor et al.
Organic & biomolecular chemistry, 8(20), 4575-4581 (2010-08-27)
A new, heterogeneous, 4 A molecular sieve-supported copper(ii) catalyst was developed and was used successfully in the A(3) coupling of alkynes, aldehydes and amines under simple reaction conditions.
Adrian Sulistio et al.
Chemical communications (Cambridge, England), 47(4), 1151-1153 (2010-12-15)
Highly functionalized water soluble core cross-linked star (CCS) polymers having degradable cores and hierarchical functionalities spanning from the peripheral groups along the arms to the core have been synthesized entirely from amino acid building blocks. The core-isolated moieties were shown
Claudio Gnaccarini et al.
Organic & biomolecular chemistry, 10(27), 5258-5265 (2012-06-02)
Transglutaminases (TGases) catalyse the transamidation of glutamine residues with primary amines. Herein we report the first FRET-based activity assay for the direct detection of the ligation (transamidation) reaction mediated by tissue TGase (TG2). This novel assay was then used in
Olga P Pereshivko et al.
Organic letters, 12(11), 2638-2641 (2010-05-06)
An efficient, microwave-assisted Cu(I)-catalyzed one-pot coupling of a ketone, an alkyne, and a primary amine (KA(2) coupling) is described, giving access to secondary propargylamines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service