Skip to Content
Merck
All Photos(1)

Key Documents

P49007

Sigma-Aldrich

Piperine

≥97%

Synonym(s):

(E,E)-5-(3,4-Methylenedioxyphenyl)-2,4-pentadienoylpiperidide, 1-Piperoylpiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO3
CAS Number:
Molecular Weight:
285.34
Beilstein:
90741
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥97%

form

crystalline
powder, crystals or chunks
solid

mp

131-135 °C (lit.)

SMILES string

O=C(\C=C\C=C\c1ccc2OCOc2c1)N3CCCCC3

InChI

1S/C17H19NO3/c19-17(18-10-4-1-5-11-18)7-3-2-6-14-8-9-15-16(12-14)21-13-20-15/h2-3,6-9,12H,1,4-5,10-11,13H2/b6-2+,7-3+

InChI key

MXXWOMGUGJBKIW-YPCIICBESA-N

Gene Information

human ... CYP3A4(1576)

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for synthesis of:
  • Dimeric amide alkaloids[1]
  • Piperoyl-amino acid conjugates used for antileishmanial activity[2]
  • Piperine derived amides with TRPV1 activity[3]
  • Piperine analogues as S. aureus NorA efflux pump inhibitors[4]
  • Bioactive conjugates of curcumin with antimicrobial and antiproliferative properties[5]
  • Piperine derivatives for use as trypanocidal agents[6]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V. R. S. Rao, et al.,
Tetrahedron, 67, 1885-1892 (2011)
Jiu-Wang Yu et al.
Molecular diversity, 25(1), 233-248 (2020-03-05)
Piperine is the main active component of Piper longum L., which is also the main component of anti-sciatica Mongolian medicine Naru Sanwei pill. It has many pharmacological activities such as anti-inflammatory and immune regulation. This paper aims to preliminarily explore
Welisson da Silva Ferreira et al.
Bioorganic & medicinal chemistry, 16(6), 2984-2991 (2008-01-30)
We herein describe the synthesis and characterization of nine new 1,3,4-thiadiazolium-2-phenylamine chlorides derived from natural piperine. We evaluate their toxic effects against the different evolutive forms of Trypanosoma cruzi, and the host cell (murine macrophages). The results obtained show that
Janine Zaugg et al.
Journal of natural products, 73(2), 185-191 (2010-01-21)
A plant extract library was screened for GABA(A) receptor activity making use of a two-microelectrode voltage clamp assay on Xenopus laevis oocytes. An ethyl acetate extract of black pepper fruits [Piper nigrum L. (Piperaceae) 100 microg/mL] potentiated GABA-induced chloride currents
Clara Grosso et al.
Phytochemical analysis : PCA, 24(2), 141-147 (2012-09-19)
Depression is a mental disease causing large personal and socio-economic problems, and new improved drugs are therefore needed. Selective monoamine oxidase A (MAO-A) inhibitors are potential anti-depressants, but discovering new MAO-A inhibitors from natural sources by bioassay-guided approaches are a

Questions

  1. piperine (5g) is currently on sale. When does it expire? How long can it be stored?

    1 answer
    1. The recommended storage temperature for item P49007 is room temperature. This product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one-year warranty from the date of shipment. For more information, you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service