Skip to Content
Merck
All Photos(4)

Documents

M3302

Sigma-Aldrich

2-Mercaptobenzothiazole

97%

Synonym(s):

2-Benzothiazolethiol, MBT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NS2
CAS Number:
Molecular Weight:
167.25
Beilstein:
119484
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

177-181 °C (lit.)

SMILES string

Sc1nc2ccccc2s1

InChI

1S/C7H5NS2/c9-7-8-5-3-1-2-4-6(5)10-7/h1-4H,(H,8,9)

InChI key

YXIWHUQXZSMYRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Mercaptobenzothiazole (MBT) has been used in the synthesis of MBT functionalized mesoporous silica which can be used as an adsorbent for the removal of Hg(II) from aqueous solution.

It can also be used as a:
  • Reference compound in photocatalytic activity tests under UV or visible light irradiation.
  • Starting material for the synthesis of conjugates of 2-MBT for antitubercular activity studies.
  • Starting material for the synthesis of 4-thiazolidinones.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

392.0 °F - closed cup

Flash Point(C)

200 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of 2-mercaptobenzothiazole-derivatized mesoporous silica and removal of Hg (II) from aqueous solution
Perez-Quintanilla D, et al.
Journal of Environmental Monitoring, 8(1), 214-222 (2006)
Enhanced photocatalytic activity of Ce3+?TiO2 for 2-mercaptobenzothiazole degradation in aqueous suspension for odour control
Li FB, et al.
Applied Catalysis, 285(1-2), 181-189 (2005)
A facile microwave enhanced synthesis of sulfur-containing 5-membered heterocycles derived from 2-mercaptobenzothiazole over ZnCl2/DMF and antimicrobial activity evaluation
Desai KG, et al.
Journal of Sulfur Chemistry, 27(4), 315-328 (2006)
Kuan-Jen Chen et al.
Talanta, 81(4-5), 1670-1675 (2010-05-06)
This research reports a sensor array consisting of three types of surface-modified nanoparticles that exhibit localized surface plasmon resonance (LSPR) extinctions at different wavelength regions of a UV-vis spectrum. By simultaneously measuring the ensemble of the LSPR bands of the
Itai Chipinda et al.
Chemical research in toxicology, 20(8), 1084-1092 (2007-07-17)
The rubber accelerator, 2-mercaptobenzothiazole (MBT), has been reported to cause allergic contact dermatitis from gloves and other rubber products, but its chemical fate when exposed to occupational oxidants and the mechanism of its pathogenesis are not known. It was hypothesized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service