Skip to Content
Merck
All Photos(2)

Key Documents

M27301

Sigma-Aldrich

Methyl acrylate

99%, contains ≤100 ppm monomethyl ether hydroquinone as inhibitor

Synonym(s):

Methylacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCOOCH3
CAS Number:
Molecular Weight:
86.09
Beilstein:
605396
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

3 (vs air)

vapor pressure

67.5 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

874 °F

contains

≤100 ppm monomethyl ether hydroquinone as inhibitor

expl. lim.

14.5 %

refractive index

n20/D 1.402 (lit.)

bp

80 °C (lit.)

mp

−75 °C (lit.)

density

0.956 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(=O)C=C

InChI

1S/C4H6O2/c1-3-4(5)6-2/h3H,1H2,2H3

InChI key

BAPJBEWLBFYGME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl acrylate is an acrylic monomer. It is useful in the preparation of polymers having rigid, flexible, ionic, nonionic, hydrophobic, or hydrophilic properties. Mechanism of the copolymerization of ethylene and α-olefins with methyl acrylate has been studied. Heck reaction of iodobenzene with methyl acrylate in the presence of triethylamine and sodium carbonate has been reported. The reversible addition fragmentation chain transfer (RAFT) bulk polymerization of methyl acrylate using 1-phenylethyl dithiobenzoate and 2-(2-cyanopropyl) dithiobenzoate (RAFT agents) has been studied. Reactive intermediates formed during the Heck reaction of methyl acrylate have been characterized by NMR spectroscopy.

Application

Methyl acrylate was used in the preparation of poly(methyl acrylate) elastomers. It was also used in the synthesis of α,ω-dicarboxylic acid esters.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

27.0 °F - closed cup

Flash Point(C)

-2.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Origin of inhibition effects in the reversible addition fragmentation chain transfer (RAFT) polymerization of methyl acrylate.
Perrier S, et al.
Macromolecules, 35(22), 8300-8306 (2002)
Mechanistic studies of the palladium-catalyzed copolymerization of ethylene and a-olefins with methyl acrylate.
Mecking S, et al.
Journal of the American Chemical Society, 120(5), 888-899 (1998)
Effects of dispersion and aggregation of silica in the reinforcement of poly (methyl acrylate) elastomers.
Pu Z, et al.
Chemistry of Materials, 9(11), 2442-2447 (1997)
Rybak A and Meier MAR.
Green Chemistry, 9(12), 1356-1361 (2007)
Characterization of Reactive Intermediates in Palladium-Catalyzed Arylation of Methyl Acrylate (Heck Reaction).
Brown JM and Hii KKM.
Angewandte Chemie (International Edition in English), 35(6), 657-659 (1996)

Articles

Our strategy is to synthesize mesoporous carbonaceous materials (“Starbons”) using mesoporous expanded starch as the precursor without the need for a templating agent.

RAFT polymerization offers living characteristics to radical polymerization, contributing versatility to reversible deactivation radical polymerization methods.

RAFT polymerization offers living characteristics to radical polymerization, contributing versatility to reversible deactivation radical polymerization methods.

RAFT polymerization offers living characteristics to radical polymerization, contributing versatility to reversible deactivation radical polymerization methods.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service