Skip to Content
Merck
All Photos(1)

Documents

M1641

Sigma-Aldrich

Methoxyphenamine hydrochloride

≥99%

Synonym(s):

2-Methoxy-N,α-dimethylbenzeneethanamine hydrochloride, NSC 65644

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17NO · HCl
CAS Number:
Molecular Weight:
215.72
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

SMILES string

Cl.CNC(C)Cc1ccccc1OC

InChI

1S/C11H17NO.ClH/c1-9(12-2)8-10-6-4-5-7-11(10)13-3;/h4-7,9,12H,8H2,1-3H3;1H

InChI key

FGSJNNQVSUVTPW-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation of optical isomers of methoxyphenamine and its metabolites as N-heptafluorobutyryl-L-prolyl derivatives by fused-silica capillary gas chromatography.
S D Roy et al.
Journal of chromatography, 431(1), 210-215 (1988-09-23)
Hajime Miyaguchi et al.
Journal of chromatography. A, 1129(1), 105-110 (2006-07-29)
A microchip-based liquid-liquid extraction for the gas chromatography analysis of urine for amphetamine-type stimulants has been developed. Partially modified microchannels with the capillarity restricted modification (CARM) method were employed for stabilizing the interface consisting of 1-chlorobutane and alkalized urine. Reliability
S D Roy et al.
Clinical pharmacology and therapeutics, 38(2), 128-133 (1985-08-01)
Urine and plasma concentrations of methoxyphenamine (MP) and three of its metabolites were determined after a single oral 60.3 mg dose of MP hydrochloride to healthy subjects of known debrisoquin (D) phenotype. Urine was collected from five extensive (EM) and
Y Nakahara et al.
Forensic science international, 63(1-3), 109-119 (1993-12-01)
The excretion of methoxyphenamine (MOP) and methamphetamine (MA) into beards has been studied. Six healthy male subjects orally took 50 mg of MOP at a single dose and 7 doses for a successive 7 days. Their beard hairs were collected
Mario Thevis et al.
European journal of mass spectrometry (Chichester, England), 14(3), 145-152 (2008-08-19)
Methoxyphenamine (o-methoxy-N,alpha-dimethylphenethylamine, Orthoxine) used in earlier times as a bronchodilator is prohibited in sports according to the regulations of the World Anti-Doping Agency (WADA). The drug and several of its metabolites are commonly analysed in doping control screening assays using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service