Skip to Content
Merck
All Photos(1)

Documents

L040150

Sigma-Aldrich

Cap 1

Synonym(s):

Cap A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.21

type

for DNA synthesis

Quality Level

product line

Proligo Reagents

form

liquid

storage condition

dry at room temperature

technique(s)

DNA synthesis: suitable

impurities

≤200 ppm water content (Karl Fischer)

color

clear

suitability

conforms to structure (GC, Formulation)

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Cap 1 solution consists of n-methylimidazole, lutidine, and acetonitrile (ACN).

Application

Cap 1 is suitable for use after the phosphoramidite reaction/coupling step.

Biochem/physiol Actions

Cap 1 solution aborts chains bearing unreacted 5′hydroxyl groups due to failure of reaction with activated phosphoramidite (typically, 1 to 2% of growing oligonucleotide chains in each synthesis cycle). These unreacted 5′hydroxyl groups are capped with an acetyl group and thus rendered unreactive for subsequent synthesis steps.

Preparation Note

Acetonitrile / Lutidine / N-Methylimidazole 8/1/1 (v/v/v)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service