Skip to Content
Merck
All Photos(2)

Documents

H25808

Sigma-Aldrich

4-Benzylphenol

99%

Synonym(s):

α-Phenyl-p-cresol, 4-Hydroxydiphenylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2C6H4OH
CAS Number:
Molecular Weight:
184.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

bp

198-200 °C/10 mmHg (lit.)

mp

79-81 °C (lit.)

SMILES string

Oc1ccc(Cc2ccccc2)cc1

InChI

1S/C13H12O/c14-13-8-6-12(7-9-13)10-11-4-2-1-3-5-11/h1-9,14H,10H2

InChI key

HJSPWKGEPDZNLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanna Andersson et al.
Bioorganic & medicinal chemistry, 16(14), 6924-6935 (2008-06-17)
Analogues of the hexapeptide angiotensin IV (Ang IV, Val(1)-Tyr(2)-Ile(3)-His(4)-Pro(5)-Phe(6)) encompassing a 4-hydroxydiphenylmethane scaffold replacing Tyr(2) and a phenylacetic or benzoic acid moiety replacing His(4)-Pro(5)-Phe(6) have been synthesized and evaluated in biological assays. The analogues inhibited the proteolytic activity of cystinyl
Kei Takemoto et al.
Mutation research, 519(1-2), 199-204 (2002-08-06)
The genotoxic potential of benzophenone and its metabolically related compounds, benzhydrol and p-benzoylphenol, was investigated using human cytochrome P450 (P450) enzymes. Benzophenone and its two metabolites (0.1-1mM) showed a suppression of bacterial growth without any P450 system, but no induction
A W Stocklinski
Xenobiotica; the fate of foreign compounds in biological systems, 11(6), 425-432 (1981-06-01)
1. Urine and faeces, and two-hour bile samples from adult male rats dosed with [14C]diphenylmethane were analysed for benzhydrol and 2- and 4-hydroxydiphenyl-methane by silica gel GF t.l.c. and 14C-determination. 2. Mean values of 48.4% and 17.7% of the administered
Brandi D Sanders et al.
Bioorganic & medicinal chemistry, 17(19), 7031-7041 (2009-09-08)
The sirtuin proteins are broadly conserved NAD(+)-dependent deacetylases that are implicated in diverse biological processes including DNA recombination and repair, transcriptional silencing, longevity, apoptosis, axonal protection, insulin signaling, and fat mobilization. Because of these associations, the identification of small molecule
Peter J McNamara et al.
Antimicrobial agents and chemotherapy, 53(5), 1898-1906 (2009-02-19)
Menstrual toxic shock syndrome is a rare but potentially life-threatening illness manifest through the actions of Staphylococcus aureus toxic shock syndrome toxin 1 (TSST-1). Previous studies have shown that tampon additives can influence staphylococcal TSST-1 production. We report here on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service