Skip to Content
Merck
All Photos(2)

Key Documents

GF86850029

Tin

foil, 25x25mm, thickness 0.15mm, as rolled, 98.8%

Synonym(s):

Tin, SN000390

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sn
CAS Number:
Molecular Weight:
118.71
MDL number:
UNSPSC Code:
12141745
PubChem Substance ID:
NACRES:
NA.23

Assay

≥98.8%

form

foil

manufacturer/tradename

Goodfellow 868-500-29

resistivity

11 μΩ-cm, 20°C

bp

2270 °C (lit.)

mp

231.9 °C (lit.)

density

7.310 g/mL at 25 °C (lit.)

SMILES string

[Sn]

InChI

1S/Sn

InChI key

ATJFFYVFTNAWJD-UHFFFAOYSA-N

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrey A Gurtovenko et al.
The Journal of chemical physics, 130(21), 215107-215107 (2009-06-11)
The electrostatic properties of lipid membranes are of profound importance as they are directly associated with membrane potential and, consequently, with numerous membrane-mediated biological phenomena. Here we address a number of methodological issues related to the computation of the electrostatic
J Strang et al.
Addiction (Abingdon, England), 92(6), 673-683 (1997-06-01)
The history of heroin smoking and the subsequent development and spread of 'chasing the dragon' are examined. The first heroin smoking originated in Shanghai in the 1920s and involved use of porcelain bowls and bamboo tubes, thereafter spreading across much
K-H Utz et al.
Journal of oral rehabilitation, 29(5), 458-466 (2002-05-25)
In a dentate subject a jaw relation can either be determined in maximum intercuspation and is as such given by the occlusal morphology, or the mandibular position can be allocated according to the centric position of the condyles. For comprehensive
Jianan Liu et al.
Animal reproduction science, 134(3-4), 197-202 (2012-08-28)
Cryopreservation of ovarian tissue has been the only effective way of ex situ conservation of female germplasm in avian species. A novel needle-in-straw (NIS) vitrification method was developed to store tissue in straws instead of cryovials. Fragments of ovarian tissue
Inorganic tin: chemistry, disposition and role in nuclear medicine diagnostic skeletal imaging agents.
M D Francis et al.
International journal of nuclear medicine and biology, 8(2-3), 145-152 (1981-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service