GF62961486
Brass alloy, Cu63Zn37
foil, 100 x 100mm, 0.0125mm thickness, half hard, light tight (lt)
Synonym(s):
CU020210
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
light tight (lt)
form
foil (half hard)
manufacturer/tradename
Goodfellow 629-614-86
size × thickness
100 mm × 100 mm × 0.0125 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service